Learn More
Aviva Systems Biology Corp CLSTN3 Antibody - N-terminal region (ARP47060_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47060_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CLSTN3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CLSTN3. Alternate names: CSTN3, KIAA0726, MGC131797, MGC138488, alcbeta, CDHR14ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62837