Learn More
Aviva Systems Biology Corp CLTA Antibody - C-terminal region (ARP59458_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59458_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CLTA. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human CLTA. Alternate names: LCAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-703