Learn More
enQuire Bioreagents Clathrin, Light Chain anti-Human Monoclonal [SPM174], Size: 100ug
Supplier:  enQuire Bioreagents 1211MSM1XP1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: SPM174. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b kappa. Positive Control: HeLa cells. Placenta or Prostate Carcinoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Purified recombinant N-terminal fragment of human Clathrin Light Chain. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 31-44kDa.Chromosome: 9p13.3. Gene Symbol: CLTA and CLTB. Gene ID: 1212. Ensemble: ENSG00000175416. Uniprot: XP_016864515.
Catalog No. 50-188-4613