Learn More
Ibidi USA Culture-Insert 2 Well 24, #1.5 polymer coverslip
SDP
Catalog No. NC2124782
Encompass
Complete solution for wound healing experiments, requiring only a few steps from sample preparation to image analysis.Reproducible experiments owing to: a defined 500 µm cell free gap, no leakage during cultivation, and no material being left behind after the insert’s removal. For screening applications. High parallelization. Based on µ-Plate 24 Well Black ID 14 mm – i.e., suitable for fluorescence and high resolution microscopy.
This item is not returnable. View return policy
Catalog No. NC2124782 Supplier Ibidi USA Supplier No. 80242
Complete solution for wound healing experiments, requiring only a few steps from sample preparation to image analysis.Reproducible experiments owing to: a defined 500 µm cell free gap, no leakage during cultivation, and no material being left behind after the insert’s removal. For screening applications. High parallelization. Based on µ-Plate 24 Well Black ID 14 mm – i.e., suitable for fluorescence and high resolution microscopy.