Learn More
Aviva Systems Biology Corp CLU Antibody - C-terminal region (ARP61142_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61142_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CLU. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human CLU. Alternate names: AAG4, APOJ, CLI, KUB1, MGC24903, SGP-2, SGP2, SP-40, APO-J, NA1/NA2ApplicationWB, IHC. Species reactivity: Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-268