Learn More
Aviva Systems Biology Corp CLUAP1 Antibody - C-terminal region (ARP55150_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55150_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CLUAP1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human CLUAP1. Alternate names: FLJ13297, KIAA0643, FAP22ApplicationWB. Species reactivity: Cow, Goat, Horse, Human, Mouse, Pig, Rat, Yeast, Zebrafish
Catalog No. 50-286-009