Learn More
Medchemexpress LLC Clusterin/APOJ Huma 500ug | 500ug
SDP
Catalog No. 5000194915
Encompass_Preferred
Clusterin/APOJ Protein Human (HEK 293 His) expresses in HEK 293 cells with a His tag at the C-terminus Clusterin (CLU) is a multifunctional glycoprotein that has been implicated in several physiological and pathological states including Alzheimer s disease (AD)[1]
Catalog No. 50-001-94915 Supplier Medchemexpress LLC Supplier No. HYP7895500UG
Clusterin/APOJ Protein Human (HEK 293 His) expresses in HEK 293 cells with a His tag at the C-terminus Clusterin (CLU) is a multifunctional glycoprotein that has been implicated in several physiological and pathological states including Alzheimer s disease (AD)[1]