Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Clusterin (apolipoprotein J), recombinant mouse protein | 98.0% | 100UG
SDP

Catalog No. 5000176514
Encompass_Preferred

Recombinant mouse Clusterin (apolipoprotein J) produced in HEK293 cells and supplied as a C-terminal His-tagged protein for research use. The product is highly purified (~98%) and intended for biochemical and cell-based assays involving extracellular chaperone activity, protein aggregation, and binding studies. The native protein is proteolytically processed into alpha and beta chains and is glycosylated, forming a disulfide-linked heterodimer that typically migrates at ~75-80 kDa in SDS-PAGE despite a calculated precursor mass of ~51,656 Da.

  • Recombinant mouse protein expressed in HEK293 cells.
  • C-terminal His tag enables detection and purification.
  • Purity approximately 98%.
  • Available in multiple sizes for assay flexibility.
  • Stored at -20 °C for stability during long-term storage.
  • Suitable for aggregation, binding, and biomarker studies.

Catalog No. 50-001-76514 Supplier Medchemexpress LLC Supplier No. HYP70058100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse Clusterin (apolipoprotein J) produced in HEK293 cells and supplied as a C-terminal His-tagged protein for research use. The product is highly purified (~98%) and intended for biochemical and cell-based assays involving extracellular chaperone activity, protein aggregation, and binding studies. The native protein is proteolytically processed into alpha and beta chains and is glycosylated, forming a disulfide-linked heterodimer that typically migrates at ~75-80 kDa in SDS-PAGE despite a calculated precursor mass of ~51,656 Da.

  • Recombinant mouse protein expressed in HEK293 cells.
  • C-terminal His tag enables detection and purification.
  • Purity approximately 98%.
  • Available in multiple sizes for assay flexibility.
  • Stored at -20 °C for stability during long-term storage.
  • Suitable for aggregation, binding, and biomarker studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.