Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Clusterin (apolipoprotein J), recombinant mouse | >95.0% | 5UG
SDP

Catalog No. 5000176471
Encompass_Preferred

Recombinant mouse Clusterin (apolipoprotein J) is a HEK293-expressed, glycosylated protein with a C-terminal 6xHis tag that functions as an extracellular chaperone to inhibit plasma protein aggregation and amyloid fibril formation. Supplied as a lyophilized powder, it is intended for research into protein aggregation, neurodegeneration, apoptosis, and protein-protein interactions.

  • Expressed in HEK293 mammalian cells.
  • Contains a C-terminal 6xHis tag for detection and purification.
  • Glycosylated; approx. molecular weight 29-44 kDa (chains), 60-77 kDa (precursor/oligomers).
  • Purity >95.0% by reducing SDS-PAGE.
  • Endotoxin <1 EU/μg (LAL method).
  • Lyophilized formulation with 8% trehalose for stability.
  • Store at -20°C; reconstitute to ≥100 μg/mL and aliquot for long-term storage.

Catalog No. 50-001-76471 Supplier Medchemexpress LLC Supplier No. HYP700585UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse Clusterin (apolipoprotein J) is a HEK293-expressed, glycosylated protein with a C-terminal 6xHis tag that functions as an extracellular chaperone to inhibit plasma protein aggregation and amyloid fibril formation. Supplied as a lyophilized powder, it is intended for research into protein aggregation, neurodegeneration, apoptosis, and protein-protein interactions.

  • Expressed in HEK293 mammalian cells.
  • Contains a C-terminal 6xHis tag for detection and purification.
  • Glycosylated; approx. molecular weight 29-44 kDa (chains), 60-77 kDa (precursor/oligomers).
  • Purity >95.0% by reducing SDS-PAGE.
  • Endotoxin <1 EU/μg (LAL method).
  • Lyophilized formulation with 8% trehalose for stability.
  • Store at -20°C; reconstitute to ≥100 μg/mL and aliquot for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.