Learn More
Sino Biological Clusterin / Apolipoprotein J / Apo-J / CLU Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 50485RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Clusterin / Apolipoprotein J / Apo-J / CLU (rM Clusterin / Apolipoprotein J / Apo-J / CLU; Catalog#50485-M08H; NP_038520.2; Met 1-Glu 448). Clusterin / Apolipoprotein J / Apo-J / CLU specific IgG was purified by Mouse Clusterin / Apolipoprotein J / Apo-J / CLU affinity chromatography.
Catalog No. 50-224-8342