Learn More
Medchemexpress LLC Clusterin (apolipoprotein J), human recombinant (HEK293, Fc-His) | 97.4% | 5 UG
SDP
Catalog No. 5000291301
Encompass_Preferred
Recombinant human Clusterin (Apolipoprotein J) produced in HEK293 cells with an Fc-His fusion tag. Supplied as a small research quantity (5 μg) and characterized by reducing SDS-PAGE; the certificate of analysis reports 97.43% purity. Intended for biochemical assays, receptor-binding studies, and cell-based uptake/endocytosis experiments. Store and handle according to the manufacturer's instructions.
Catalog No. 50-002-91301 Supplier Medchemexpress LLC Supplier No. HYP75355UG
Recombinant human Clusterin (Apolipoprotein J) produced in HEK293 cells with an Fc-His fusion tag. Supplied as a small research quantity (5 μg) and characterized by reducing SDS-PAGE; the certificate of analysis reports 97.43% purity. Intended for biochemical assays, receptor-binding studies, and cell-based uptake/endocytosis experiments. Store and handle according to the manufacturer's instructions.