Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Clusterin (apolipoprotein J), human recombinant protein (HEK293, Fc-His) | >90.0% | 100 UG
SDP

Catalog No. 5000291285
Encompass_Preferred

Recombinant human clusterin (apolipoprotein J) expressed in HEK293 cells and supplied lyophilized with Fc and His tags. The product corresponds to UniProt P10909-1 (D23-E449) and is intended for laboratory research use only; not for clinical or human use.

  • Expressed in HEK293 cells.
  • Contains Fc fragment and C-terminal His tag.
  • Amino acid range D23-E449 (UniProt P10909-1).
  • Purity greater than 90% by reducing SDS-PAGE.
  • Lyophilized formulation from PBS/20 mM PB with trehalose and mannitol.
  • Store at -20°C; stable 2 years from receipt; aliquot and freeze for long term.
  • Do not reconstitute below 100 μg/mL in ddH2O; add carrier protein for long-term storage.

Catalog No. 50-002-91285 Supplier Medchemexpress LLC Supplier No. HYP7535100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human clusterin (apolipoprotein J) expressed in HEK293 cells and supplied lyophilized with Fc and His tags. The product corresponds to UniProt P10909-1 (D23-E449) and is intended for laboratory research use only; not for clinical or human use.

  • Expressed in HEK293 cells.
  • Contains Fc fragment and C-terminal His tag.
  • Amino acid range D23-E449 (UniProt P10909-1).
  • Purity greater than 90% by reducing SDS-PAGE.
  • Lyophilized formulation from PBS/20 mM PB with trehalose and mannitol.
  • Store at -20°C; stable 2 years from receipt; aliquot and freeze for long term.
  • Do not reconstitute below 100 μg/mL in ddH2O; add carrier protein for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.