Learn More
Enzo Life Sciences Clusterin (secretory form) (human), (recombinant) (His-tag) (10µg)
SDP
Supplier:  Enzo Life Sciences ALX2017610010
Encompass_Preferred
Alternative name: TRPM-2, Apolipoprotein J, APO-J, CLI, CLU, SGP-2. Endotoxin: < 0.1ng/µg (1 IEU/µg). Purity: ≥95% (SEC-HPLC and reducing SDS-PAGE). Formulation: Lyophilized. From a 0.2 μM filtered solution of 20mM PBS, pH 7.4 and 150mM sodium chloride, pH 7.4. Source: Secreted protein produced by transfected human cells. Human clusterin (aa 23-422) is fused at the C-terminus to a His-tag. Long Term Storage: -20°C.
Catalog No. 50-200-7983