Learn More
Enzo Life Sciences Clusterin (secretory form) (human), (recombinant) (His-tag) (50µg)
SDP
Supplier:  Enzo Life Sciences ALX2017610050
Encompass_Preferred
Alternative name: TRPM-2, Apolipoprotein J, APO-J, CLI, CLU, SGP-2. Endotoxin: < 0.1ng/µg (1 IEU/µg). Purity: ≥95% (SEC-HPLC and reducing SDS-PAGE). Formulation: Lyophilized. From a 0.2 μM filtered solution of 20mM PBS, pH 7.4 and 150mM sodium chloride, pH 7.4. Source: Secreted protein produced by transfected human cells. Human clusterin (aa 23-422) is fused at the C-terminus to a His-tag. Long Term Storage: -20°C. Use/Stability: Lyophilized protein should be stored at < -20°C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7°C for 2-7 days. Working aliquots are stable for up to 3 months when stored at -20°C. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -20°C.
Catalog No. 50-200-7984