Learn More
Enzo Life Sciences Clusterin (human) monoclonal antibody (CLI-9) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804126C100
Encompass_Preferred
Clone: CLI-9. Isotype: IgG1. Immunogen: Purified human clusterin (TRPM-2). Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human. Applications: ICC, IHC (FS), IHC (PS), IP, WB. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-9787