Learn More
Cayman Chemical ClusterIn MoncLNalCLN 9 100uL
SDP
Catalog No. 502914924
Encompass_Preferred
An inhibitor of cardiac myosin ATPase (IC50 1.26 UM for the bovine enzyme) reduces fractional shortening in isolated rat cardiac ventricular myocytes (IC50 7.9 UM) as well as in rats in a dose-dependent manner
Catalog No. 50-291-4924 Supplier Cayman Chemical Supplier No. 37066100 UL
An inhibitor of cardiac myosin ATPase (IC50 1.26 UM for the bovine enzyme) reduces fractional shortening in isolated rat cardiac ventricular myocytes (IC50 7.9 UM) as well as in rats in a dose-dependent manner