Learn More
Cayman Chemical ClusterIn MoncLNalCLN 9 200uL
SDP
Catalog No. 502914926
Encompass_Preferred
An inhibitor of cardiac myosin ATPase (IC50 1.26 UM for the bovine enzyme) reduces fractional shortening in isolated rat cardiac ventricular myocytes (IC50 7.9 UM) as well as in rats in a dose-dependent manner
Catalog No. 50-291-4926 Supplier Cayman Chemical Supplier No. 37066200 UL
An inhibitor of cardiac myosin ATPase (IC50 1.26 UM for the bovine enzyme) reduces fractional shortening in isolated rat cardiac ventricular myocytes (IC50 7.9 UM) as well as in rats in a dose-dependent manner