Learn More
AdipoGen Clusterin NF ms recHis
SDP
Catalog No. 502563032
Encompass_Preferred
Protein. Mouse nuclear clusterin aa 22-448 is fused at the C-terminus to a His-tag. Source E. coli. Endotoxin content <1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in 55mM TRIS-Cl, pH 8.2, containing 150mM NaCl. Purity >90% SDS-PAGE. Clusterin shares homology with the small heat shock protein family of molecular chaperones. The mature secreted form of the protein is a glycosylated, 80-kDa disulfide-linked heterodimer of alpha and beta subunits produced by internal cleavage. Clusterin is expressed in virtually all tissues and found in all human fluids. It is involved in numerous physiological processes important for carcinogenesis and tumor growth, including apoptotic cell death, cell cycle regulation, DNA repair, cell adhesion, tissue remodeling, lipid transportation, membrane recycling, and immune system regulation. Clusterin also exists as a nuclear protein.
Catalog No. 50-256-3032 Supplier AdipoGen Supplier No. AG40A0057C050
Protein. Mouse nuclear clusterin aa 22-448 is fused at the C-terminus to a His-tag. Source E. coli. Endotoxin content <1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in 55mM TRIS-Cl, pH 8.2, containing 150mM NaCl. Purity >90% SDS-PAGE. Clusterin shares homology with the small heat shock protein family of molecular chaperones. The mature secreted form of the protein is a glycosylated, 80-kDa disulfide-linked heterodimer of alpha and beta subunits produced by internal cleavage. Clusterin is expressed in virtually all tissues and found in all human fluids. It is involved in numerous physiological processes important for carcinogenesis and tumor growth, including apoptotic cell death, cell cycle regulation, DNA repair, cell adhesion, tissue remodeling, lipid transportation, membrane recycling, and immune system regulation. Clusterin also exists as a nuclear protein.