Learn More
Alpco Ltd Rat Clusterin ELISA
SDP
Supplier:  Alpco Ltd 41CLURTE01
Encompass
The Rat Clusterin ELISA is a competitive enzyme-linked immunosorbent assay for the quantitative determination of clusterin in rat serum, plasma, or urine.
Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-751-3572