Learn More
AdipoGen Clusterin SF hrec
SDP
Catalog No. 502563038
Encompass_Preferred
Protein. Signal peptide sequence and the human secretory clusterin aa 1-449 are fused at the C-terminus to a FLAG -tag. Source HEK 293 cells. Endotoxin content <0.01EU/µg purified protein LAL test Lonza. Lyophilized. Contains PBS. Purity >90% SDS-PAGE. Clusterin shares homology with the small heat shock protein family of molecular chaperones. The mature secreted form of the protein is a glycosylated, 80kDa disulfide-linked heterodimer of alpha and beta subunits produced by internal cleavage. Clusterin is expressed in virtually all tissues and found in all human fluids. It is involved in numerous physiological processes important for carcinogenesis and tumor growth, including apoptotic cell death, cell cycle regulation, DNA repair, cell adhesion, tissue remodeling, lipid transportation, membrane recycling and immune system regulation. Clusterin also exists as a nuclear protein.
Catalog No. 50-256-3038 Supplier AdipoGen Supplier No. AG40A0050YC010
Protein. Signal peptide sequence and the human secretory clusterin aa 1-449 are fused at the C-terminus to a FLAG -tag. Source HEK 293 cells. Endotoxin content <0.01EU/µg purified protein LAL test Lonza. Lyophilized. Contains PBS. Purity >90% SDS-PAGE. Clusterin shares homology with the small heat shock protein family of molecular chaperones. The mature secreted form of the protein is a glycosylated, 80kDa disulfide-linked heterodimer of alpha and beta subunits produced by internal cleavage. Clusterin is expressed in virtually all tissues and found in all human fluids. It is involved in numerous physiological processes important for carcinogenesis and tumor growth, including apoptotic cell death, cell cycle regulation, DNA repair, cell adhesion, tissue remodeling, lipid transportation, membrane recycling and immune system regulation. Clusterin also exists as a nuclear protein.