Learn More
Research Products International Corp ClusterTubes, Non-RKed, 8 Tube Strip, 125 Strips
SDP
Catalog No. 502229662
Encompass
ClusterTubes are ideal for dilution, mixing, harvesting, culture assays, screening and storage applications. Rack systems are also suitable as a reaction vessel in robotic workstations or as a long-term storage vessel in sample library. Tube are available as individual tubes, strips of 8 tubes or 12 tubes. Racks feature bold, black alphanumeric indexing and are made from autoclavable virgin polyproplyene; caps are polyethylene material; and racked tubes are supplied with clear lid. Tube volume is 1.2ml.
DNase, RNase, DNA, PCR inhibitor & pyrogen free
Ideal for Automation, HTS
SBS footprint; 8 x 12 format
Catalog No. 50-222-9662 Supplier Research Products International Corp Supplier No. 162528
ClusterTubes are ideal for dilution, mixing, harvesting, culture assays, screening and storage applications. Rack systems are also suitable as a reaction vessel in robotic workstations or as a long-term storage vessel in sample library. Tube are available as individual tubes, strips of 8 tubes or 12 tubes. Racks feature bold, black alphanumeric indexing and are made from autoclavable virgin polyproplyene; caps are polyethylene material; and racked tubes are supplied with clear lid. Tube volume is 1.2ml.
DNase, RNase, DNA, PCR inhibitor & pyrogen free
Ideal for Automation, HTS
SBS footprint; 8 x 12 format