Learn More
Cayman Chemical ClusTRInCTRm RabbIt Mon 100uL
SDP
Catalog No. 502909969
Encompass_Preferred
An ionizable cationic lipid (pKa 6 09) has been used in the generation of LNPs for the delivery of mRNA siRNA and plasmid DNA in vitro and in vivo LNPs containing ALC-0315 and encapsulating siRNA targeting Factor VII mRNA decrease Factor VII mRNA levels in hepatocytes in mice administration of SARS-CoV-2 mRNA in ALC-0315-containing LNPs induces the production of IgG that binds to the SARS-CoV-2 BD) in rhesus macaques with a boost in antigen-specific IgG GMTs seven and 14 days after a second dose LNPs containing ALC-0315 and encapsulating ovalbumin mRNA reduce tumor volume in an ovalbumin-expressing B16/F10 murine melanoma model
Catalog No. 50-290-9969 Supplier Cayman Chemical Supplier No. 35616100 UL
An ionizable cationic lipid (pKa 6 09) has been used in the generation of LNPs for the delivery of mRNA siRNA and plasmid DNA in vitro and in vivo LNPs containing ALC-0315 and encapsulating siRNA targeting Factor VII mRNA decrease Factor VII mRNA levels in hepatocytes in mice administration of SARS-CoV-2 mRNA in ALC-0315-containing LNPs induces the production of IgG that binds to the SARS-CoV-2 BD) in rhesus macaques with a boost in antigen-specific IgG GMTs seven and 14 days after a second dose LNPs containing ALC-0315 and encapsulating ovalbumin mRNA reduce tumor volume in an ovalbumin-expressing B16/F10 murine melanoma model