Learn More
Miele, Inc. CM/1 Conductivity meter for me
Catalog No. 502870709
Encompass_Preferred
CM/1 Conductivity meter for measuring conductivity level (0-199µS/cm)
Catalog No. 50-287-0709 Supplier Miele, Inc. Supplier No. 10889920
CM/1 Conductivity meter for measuring conductivity level (0-199µS/cm)