Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cm-1758 | 2256079-39-5 | 98.4% | 532.59 g/mol | C28H32N6O5 | 10 MG
SDP

Catalog No. 5000288749
Encompass_Preferred

CM-1758 is a small-molecule histone deacetylase (HDAC) inhibitor for laboratory research; it has reported in vivo antitumor activity and induces acetylation of non-histone proteins in acute myeloid leukemia cells.

  • Histone deacetylase (HDAC) inhibitor with reported in vivo antitumor activity.
  • Induces acetylation of non-histone proteins in acute myeloid leukemia cells.
  • Molecular weight 532.59 g/mol and formula C28H32N6O5.
  • High purity (98.44% by HPLC) suitable for research applications.
  • Soluble in DMSO (100 mg/mL) for in vitro use; formulations available for in vivo dosing.
  • Powder storage: -20°C (3 years) or 4°C (2 years); follow recommended storage for solutions.

Catalog No. 50-002-88749 Supplier Medchemexpress LLC Supplier No. HY16114910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CM-1758 is a small-molecule histone deacetylase (HDAC) inhibitor for laboratory research; it has reported in vivo antitumor activity and induces acetylation of non-histone proteins in acute myeloid leukemia cells.

  • Histone deacetylase (HDAC) inhibitor with reported in vivo antitumor activity.
  • Induces acetylation of non-histone proteins in acute myeloid leukemia cells.
  • Molecular weight 532.59 g/mol and formula C28H32N6O5.
  • High purity (98.44% by HPLC) suitable for research applications.
  • Soluble in DMSO (100 mg/mL) for in vitro use; formulations available for in vivo dosing.
  • Powder storage: -20°C (3 years) or 4°C (2 years); follow recommended storage for solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.