Learn More
Waters Corp CM-30S Performance Maintenance Kit, Annual, for UPLC Systems
Catalog No. 502390688 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters Performance Maintenance Kit for the ACQUITY CM-S contains all the necessary parts to keep the instrument operating at peak performance on an annual basis.
  • Maintains instruments operating at peak performance.
  • Recommended for annual replacement.
  • Compatible with UPLC systems, including Arc Premier and UPLC H-Class systems.
  • Includes critical components for the UPLC QSM.
  • Contains one air filter.
  • Supplied as one unit per package.
Catalog No. 50-239-0688 Supplier Waters Corp Supplier No. 201000288
The Waters Performance Maintenance Kit for the ACQUITY CM-S contains all the necessary parts to keep the instrument operating at peak performance on an annual basis.
  • Maintains instruments operating at peak performance.
  • Recommended for annual replacement.
  • Compatible with UPLC systems, including Arc Premier and UPLC H-Class systems.
  • Includes critical components for the UPLC QSM.
  • Contains one air filter.
  • Supplied as one unit per package.