Learn More
TARGETMOL CHEMICALS INC CM-579 10MG
SDP
Catalog No. 502581646
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CM-579 is a dual inhibitor of G9a and DNMT( IC5016 nM 32 nM for G9a and DNMT). It has potent in vitro cellular activity in a wide range of cancer cells. purity: 99%
Catalog No. 50-258-1646 Supplier TARGETMOL CHEMICALS INC Supplier No. T10840L10MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CM-579 is a dual inhibitor of G9a and DNMT( IC5016 nM 32 nM for G9a and DNMT). It has potent in vitro cellular activity in a wide range of cancer cells. purity: 99%