Learn More
Medchemexpress LLC CM-579 trihydrochloride | 2448471-08-5 | 97.0% | 602.04 g/mol | C29H43Cl3N4O3 | 10 MG
SDP
Catalog No. 501969474
Encompass_Preferred
CM-579 trihydrochloride is a reversible, dual inhibitor of histone methyltransferase G9a and DNA methyltransferases (DNMT) with potent in vitro activity in cancer cell lines. It is supplied as a research reagent for biochemical and cellular studies and is for research use only.
  • Reversible dual inhibition of G9a and DNMT.
  • Low nanomolar potency (IC50 ≈ 16 nM for G9a, ≈ 32 nM for DNMT).
  • High reported purity (97.0%).
  • Supplied in a small-mass vial suitable for biochemical and cellular assays.
Catalog No. 50-196-9474 Supplier Medchemexpress LLC Supplier No. HY117421A10MG
CM-579 trihydrochloride is a reversible, dual inhibitor of histone methyltransferase G9a and DNA methyltransferases (DNMT) with potent in vitro activity in cancer cell lines. It is supplied as a research reagent for biochemical and cellular studies and is for research use only.
  • Reversible dual inhibition of G9a and DNMT.
  • Low nanomolar potency (IC50 ≈ 16 nM for G9a, ≈ 32 nM for DNMT).
  • High reported purity (97.0%).
  • Supplied in a small-mass vial suitable for biochemical and cellular assays.