Learn More
Medchemexpress LLC HY-117421A 5mg Medchemexpress, CM-579 trihydrochloride CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY117421A5MG
Encompass_Preferred
Medchemexpress, HY-117421A 5mg CM-579 trihydrochloride CAS: CM-579 trihydrochloride is a first-in-class reversible, dual inhibitor of G9a and DNMT , with IC 50 values of 16 nM, 32 nM for G9a and DNMT, respectively. Has potent in vitro cellular activity in a wide range of cancer cells [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2521