Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cm-579 trihydrochloride | 2448471-08-5 | 97.0% | 602.04 | C29H43Cl3N4O3 | 1 ML
SDP

Catalog No. 5000228665
Encompass_Preferred

CM-579 trihydrochloride is a reversible, dual inhibitor of histone methyltransferase G9a and DNA methyltransferases (DNMT) with low-nanomolar biochemical potency and demonstrated cellular activity. The compound is provided as a concentrated solution for in vitro studies and is used primarily in epigenetic and cancer research.

  • Dual G9a and DNMT inhibition with IC50 ≈ 16 nM (G9a) and 32 nM (DNMT).
  • Supplied as a 10 mM solution in DMSO (1 mL) and as solid quantities for flexibility.
  • Molecular weight 602.04 and CAS 2448471-08-5 for unambiguous identification.
  • Purity typically 97.0% by supplier specification.
  • Suitable for epigenetic modulation, target validation, and cancer cell studies.

Catalog No. 50-002-28665 Supplier Medchemexpress LLC Supplier No. HY117421A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

CM-579 trihydrochloride is a reversible, dual inhibitor of histone methyltransferase G9a and DNA methyltransferases (DNMT) with low-nanomolar biochemical potency and demonstrated cellular activity. The compound is provided as a concentrated solution for in vitro studies and is used primarily in epigenetic and cancer research.

  • Dual G9a and DNMT inhibition with IC50 ≈ 16 nM (G9a) and 32 nM (DNMT).
  • Supplied as a 10 mM solution in DMSO (1 mL) and as solid quantities for flexibility.
  • Molecular weight 602.04 and CAS 2448471-08-5 for unambiguous identification.
  • Purity typically 97.0% by supplier specification.
  • Suitable for epigenetic modulation, target validation, and cancer cell studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.