Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CM-579 trihydrochloride | 2448471-08-5 | 97.0% | 602.04 g/mol | C29H43Cl3N4O3 | 25 MG
SDP

Catalog No. 5000228692
Encompass_Preferred

CM-579 trihydrochloride is a reversible, dual inhibitor of histone methyltransferase G9a and DNA methyltransferases (DNMT), supplied as the trihydrochloride salt for biochemical and cellular research.

  • Reversible dual inhibitor of G9a and DNMT for epigenetic research.
  • High purity (97.0%) appropriate for experimental assays.
  • Molecular weight 602.04 g/mol and formula C29H43Cl3N4O3 for compound identification.
  • Available in small milligram pack sizes for compound screening and dose-response studies.
  • Offered as a DMSO solution option for convenient in vitro use.

Catalog No. 50-002-28692 Supplier Medchemexpress LLC Supplier No. HY117421A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CM-579 trihydrochloride is a reversible, dual inhibitor of histone methyltransferase G9a and DNA methyltransferases (DNMT), supplied as the trihydrochloride salt for biochemical and cellular research.

  • Reversible dual inhibitor of G9a and DNMT for epigenetic research.
  • High purity (97.0%) appropriate for experimental assays.
  • Molecular weight 602.04 g/mol and formula C29H43Cl3N4O3 for compound identification.
  • Available in small milligram pack sizes for compound screening and dose-response studies.
  • Offered as a DMSO solution option for convenient in vitro use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.