Learn More
Medchemexpress LLC CM-579 trihydrochloride | 2448471-08-5 | MFCD07996200 | 97.0% | 602.04 | C29H43Cl3N4O3 | 50 MG
SDP
Catalog No. 5000228664
Encompass_Preferred
CM-579 trihydrochloride is a research reagent that functions as a reversible, dual inhibitor of the histone methyltransferase G9a and DNA methyltransferases (DNMT). Supplied as the trihydrochloride salt at 97.0% purity and a molecular weight of 602.04, it is intended for in vitro biochemical and cellular epigenetic studies.
  • Reversible, dual inhibition of G9a and DNMT.
  • High purity (97.0%) suitable for research assays.
  • Supplied as a trihydrochloride salt for improved stability.
  • Available in small-scale quantities for screening, for example 50 MG.
  • Suitable for biochemical and cellular assay applications.
Catalog No. 50-002-28664 Supplier Medchemexpress LLC Supplier No. HY117421A50MG
CM-579 trihydrochloride is a research reagent that functions as a reversible, dual inhibitor of the histone methyltransferase G9a and DNA methyltransferases (DNMT). Supplied as the trihydrochloride salt at 97.0% purity and a molecular weight of 602.04, it is intended for in vitro biochemical and cellular epigenetic studies.
  • Reversible, dual inhibition of G9a and DNMT.
  • High purity (97.0%) suitable for research assays.
  • Supplied as a trihydrochloride salt for improved stability.
  • Available in small-scale quantities for screening, for example 50 MG.
  • Suitable for biochemical and cellular assay applications.