Learn More
MSC CM Anchor Shackle, 17000 Lb WLL, Screw Pin, Carbon Steel
Catalog No. 5000513465
Encompass
This anchor shackle features a screw pin and is made from carbon steel with a powder coated finish, suitable for applications requiring a working load limit of 17000 Lb.
  • 17000 lb working load limit.
  • Anchor shackle type.
  • Screw pin type.
  • Carbon steel material.
  • 3.31 in jaw inside height.
  • 1.44 in inside width.
  • 2.14 in overall thickness.
  • Powder coated finish.
  • 0.875 in head/holder diameter.
  • Meets ASME B30.26 standards.
Catalog No. 50-005-13465 Supplier MSC Supplier No. 10248060
This anchor shackle features a screw pin and is made from carbon steel with a powder coated finish, suitable for applications requiring a working load limit of 17000 Lb.
  • 17000 lb working load limit.
  • Anchor shackle type.
  • Screw pin type.
  • Carbon steel material.
  • 3.31 in jaw inside height.
  • 1.44 in inside width.
  • 2.14 in overall thickness.
  • Powder coated finish.
  • 0.875 in head/holder diameter.
  • Meets ASME B30.26 standards.