Learn More
MSC CM Anchor Shackle, Round Pin, 5/16 inch, 1500 lb WLL, Carbon Steel, Galvanized
Catalog No. 5000530604
Encompass
The product is a carbon anchor shackle with a round pin and a galvanized finish, designed for reliable performance in various lifting and rigging applications. It is manufactured from technically advanced domestic micro alloy steel with optimal hardness for strength and ductility, ensuring durability and safety.Key features and benefits include:
  • Manufactured from technically advanced domestic micro alloy steel.
  • Pins are alloy quenched and tempered.
  • Working load limit and traceability codes are permanently marked on the body.
  • Designed to show major deformation before failure, enhancing safety.
  • Meets dimensional, marking, and performance requirements of Federal Specification RR-C-271.
  • Galvanized finish meets ASTM A153.
  • Features a 6:1 design factor.
  • Can be side loaded or used for multiple connections.
Catalog No. 50-005-30604 Supplier MSC Supplier No. 64618366
The product is a carbon anchor shackle with a round pin and a galvanized finish, designed for reliable performance in various lifting and rigging applications. It is manufactured from technically advanced domestic micro alloy steel with optimal hardness for strength and ductility, ensuring durability and safety.Key features and benefits include:
  • Manufactured from technically advanced domestic micro alloy steel.
  • Pins are alloy quenched and tempered.
  • Working load limit and traceability codes are permanently marked on the body.
  • Designed to show major deformation before failure, enhancing safety.
  • Meets dimensional, marking, and performance requirements of Federal Specification RR-C-271.
  • Galvanized finish meets ASTM A153.
  • Features a 6:1 design factor.
  • Can be side loaded or used for multiple connections.