Learn More
MSC CM Anchor Shackle, Screw Pin, 1500 lb, Carbon Steel, Galvanized
Catalog No. 5000535016
Encompass
The CM Anchor Shackle is designed with a screw pin, offering a working load limit of 1500 lb. It is constructed from carbon steel and features a galvanized finish for enhanced durability. This shackle is suitable for various lifting and rigging applications.
  • Anchor shackle type
  • Screw pin design
  • 1500 lb load capacity
  • Carbon steel material
  • Galvanized finish
  • Meets ASTM A153 standards
  • Country of origin: United States
Catalog No. 50-005-35016 Supplier MSC Supplier No. 64313877
The CM Anchor Shackle is designed with a screw pin, offering a working load limit of 1500 lb. It is constructed from carbon steel and features a galvanized finish for enhanced durability. This shackle is suitable for various lifting and rigging applications.
  • Anchor shackle type
  • Screw pin design
  • 1500 lb load capacity
  • Carbon steel material
  • Galvanized finish
  • Meets ASTM A153 standards
  • Country of origin: United States