Learn More
MSC CM Anchor Shackle, Screw Pin, 2000 Lb Working Load Limit, Carbon Steel
Catalog No. 5000528096
Encompass
This anchor shackle features a screw pin and has a working load limit of 2000 Lb. It is constructed from carbon steel and has a painted finish. The shackle is designed with a jaw inside height of 1.2500 in and an overall thickness of 0.8400 in. It complies with ASME B30.26 standards.
  • Working load limit of 2000 Lb
  • Anchor shackle type
  • Screw pin design
  • Constructed from carbon steel
  • Jaw inside height of 1.2500 in
  • Overall thickness of 0.8400 in
  • Painted finish
  • Complies with ASME B30.26 standards
  • Manufactured in the United States
Catalog No. 50-005-28096 Supplier MSC Supplier No. 64618697
This anchor shackle features a screw pin and has a working load limit of 2000 Lb. It is constructed from carbon steel and has a painted finish. The shackle is designed with a jaw inside height of 1.2500 in and an overall thickness of 0.8400 in. It complies with ASME B30.26 standards.
  • Working load limit of 2000 Lb
  • Anchor shackle type
  • Screw pin design
  • Constructed from carbon steel
  • Jaw inside height of 1.2500 in
  • Overall thickness of 0.8400 in
  • Painted finish
  • Complies with ASME B30.26 standards
  • Manufactured in the United States