Learn More
MSC CM Chain Shackle, Screw Pin, 10000 lb WLL, Alloy Steel
Catalog No. 5000537615
Encompass
This chain shackle is designed for various lifting and rigging applications. It features a screw pin, a 10000 lb working load limit, and is constructed from durable alloy steel. This shackle is suitable for use in construction and meets ASME B30.26 standards, ensuring reliability and safety.
  • Chain shackle type
  • Screw pin for secure fastening
  • 10000 lb working load limit
  • Alloy steel construction for durability
  • Ideal for construction applications
  • 2.75 in inside width
  • 5.00 in jaw inside height
  • 1.81 in overall thickness
  • 0.75 in pin diameter
  • Powder coated finish for enhanced resistance
  • Meets ASME B30.26 standards
Catalog No. 50-005-37615 Supplier MSC Supplier No. 37314945
This chain shackle is designed for various lifting and rigging applications. It features a screw pin, a 10000 lb working load limit, and is constructed from durable alloy steel. This shackle is suitable for use in construction and meets ASME B30.26 standards, ensuring reliability and safety.
  • Chain shackle type
  • Screw pin for secure fastening
  • 10000 lb working load limit
  • Alloy steel construction for durability
  • Ideal for construction applications
  • 2.75 in inside width
  • 5.00 in jaw inside height
  • 1.81 in overall thickness
  • 0.75 in pin diameter
  • Powder coated finish for enhanced resistance
  • Meets ASME B30.26 standards