Learn More
MSC CM Clevis Hook, 1/2 in Chain Dia, Grade 100, Steel
Catalog No. 5000535536
Encompass
This Clevis Hook is designed for lifting applications, offering a high work load limit and durable construction. It features a 1/2 inch chain diameter and is made from Grade 100 steel, ensuring reliable performance in demanding environments.
  • Designed for lifting applications
  • Grade 100 steel construction
  • 1/2 inch chain diameter
  • 15,000 lb work load limit
  • Hook throat of 1.38 inches
  • Conforms to ASME B30.10 standards
  • Made in the United States
Catalog No. 50-005-35536 Supplier MSC Supplier No. 62372487
This Clevis Hook is designed for lifting applications, offering a high work load limit and durable construction. It features a 1/2 inch chain diameter and is made from Grade 100 steel, ensuring reliable performance in demanding environments.
  • Designed for lifting applications
  • Grade 100 steel construction
  • 1/2 inch chain diameter
  • 15,000 lb work load limit
  • Hook throat of 1.38 inches
  • Conforms to ASME B30.10 standards
  • Made in the United States