Learn More
MSC CM Clevis Hook, 3/8 in Chain, Grade 80, 7,100 Lbs. Load Capacity
Catalog No. 5000536324
Encompass
This clevis grab hook is engineered for demanding lifting applications. Constructed from durable alloy steel, it is compatible with 3/8 inch chain systems and provides a substantial load capacity.
  • Grade 80 chain compatibility
  • Alloy steel material
  • 7,100 Lbs. load capacity
  • 29/64 in inside diameter
  • 1/2 in pin diameter
  • 1/2 in hook throat
  • 2-5/8 in hook width
  • Made in the United States
Catalog No. 50-005-36324 Supplier MSC Supplier No. 67781542
This clevis grab hook is engineered for demanding lifting applications. Constructed from durable alloy steel, it is compatible with 3/8 inch chain systems and provides a substantial load capacity.
  • Grade 80 chain compatibility
  • Alloy steel material
  • 7,100 Lbs. load capacity
  • 29/64 in inside diameter
  • 1/2 in pin diameter
  • 1/2 in hook throat
  • 2-5/8 in hook width
  • Made in the United States