Learn More
MSC CM Electric Chain Hoist, 600 lb Working Load, 10 ft Lift
Catalog No. 5000529371
Encompass
The CM Electric Chain Hoist is designed for lifting applications, offering a 600 lb working load limit and a 10 ft lift distance. This hoist is equipped with a hook mount and operates on 115V power, providing reliable and efficient performance for various industrial tasks.
  • Hook mount for versatile installation
  • Operates on 115V for standard power compatibility
  • Lifting speed of 8
  • Motor horsepower of 0.1667
  • Meets NEMA 4 and ASME B30.16 standards for safety and performance
  • Steel hook material for durability
  • Single chain lifting mechanism
  • Manufactured in the United States
Catalog No. 50-005-29371 Supplier MSC Supplier No. 02403582
The CM Electric Chain Hoist is designed for lifting applications, offering a 600 lb working load limit and a 10 ft lift distance. This hoist is equipped with a hook mount and operates on 115V power, providing reliable and efficient performance for various industrial tasks.
  • Hook mount for versatile installation
  • Operates on 115V for standard power compatibility
  • Lifting speed of 8
  • Motor horsepower of 0.1667
  • Meets NEMA 4 and ASME B30.16 standards for safety and performance
  • Steel hook material for durability
  • Single chain lifting mechanism
  • Manufactured in the United States