Learn More
Teknova CM Galactose Broth minus Leucine, 1000mL, Sterile. 2 pack.
SDP
Catalog No. NC3309245
Encompass
Complete Minimal (CM) media is a defined medium that is an optimized mixture of amino acids, carbohydrates, vitamins and minerals. It is used to support the growth of eukaryotic cells, making it ideal for cell-based experiments.
Catalog No. NC3309245 Supplier Teknova Supplier No. C91102PK
Complete Minimal (CM) media is a defined medium that is an optimized mixture of amino acids, carbohydrates, vitamins and minerals. It is used to support the growth of eukaryotic cells, making it ideal for cell-based experiments.