Learn More
Teknova CM Galactose Broth minus Tryptophan and Uracil. 1000mL, Sterile.
SDP
Catalog No. 502388174
Encompass
Complete Minimal (CM) media is a defined medium that is an optimized mixture of amino acids, carbohydrates, vitamins and minerals. It is used to support the growth of eukaryotic cells, making it ideal for cell-based experiments.
Catalog No. 50-238-8174 Supplier Teknova Supplier No. C9130
Complete Minimal (CM) media is a defined medium that is an optimized mixture of amino acids, carbohydrates, vitamins and minerals. It is used to support the growth of eukaryotic cells, making it ideal for cell-based experiments.