Learn More
Teknova CM Galactose Broth minus Tryptophan. 1000mL, Sterile. 2 Pack.
SDP
Catalog No. 502387872
Encompass
Complete Minimal (CM) media is a defined medium that is an optimized mixture of amino acids, carbohydrates, vitamins and minerals. It is used to support the growth of eukaryotic cells, making it ideal for cell-based experiments.
Catalog No. 50-238-7872 Supplier Teknova Supplier No. C91152PK
Complete Minimal (CM) media is a defined medium that is an optimized mixture of amino acids, carbohydrates, vitamins and minerals. It is used to support the growth of eukaryotic cells, making it ideal for cell-based experiments.