Learn More
Teknova CM Glucose Agar Plates minus Histidine. 100mm Plates, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387854
Encompass
Complete Minimal (CM) media is a defined medium that is an optimized mixture of amino acids, carbohydrates, vitamins and minerals. It is used to support the growth of eukaryotic cells, making it ideal for cell-based experiments.
Catalog No. 50-238-7854 Supplier Teknova Supplier No. C37904PK
Complete Minimal (CM) media is a defined medium that is an optimized mixture of amino acids, carbohydrates, vitamins and minerals. It is used to support the growth of eukaryotic cells, making it ideal for cell-based experiments.