Learn More
Teknova CM Glucose Agar Plates minus Tryptophan and Uracil. 100mm, 20 Plates/Sleeve, Sterile.
SDP
Catalog No. 502388013
Encompass
Complete Minimal (CM) media is a defined medium that is an optimized mixture of amino acids, carbohydrates, vitamins and minerals. It is used to support the growth of eukaryotic cells, making it ideal for cell-based experiments.
Catalog No. 50-238-8013 Supplier Teknova Supplier No. C3260
Complete Minimal (CM) media is a defined medium that is an optimized mixture of amino acids, carbohydrates, vitamins and minerals. It is used to support the growth of eukaryotic cells, making it ideal for cell-based experiments.