Learn More
Teknova CM Glucose Broth minus Leucine, 1000mL, Sterile.
SDP
Supplier:  Teknova C8120
Encompass
Complete Minimal (CM) media is a defined medium that is an optimized mixture of amino acids, carbohydrates, vitamins and minerals. It is used to support the growth of eukaryotic cells, making it ideal for cell-based experiments.
Catalog No. NC3309241