Learn More
MSC CM Manual Hand Chain Hoist, 10000 lb, 20 ft Lift, 2 Chain
Catalog No. 5000529849
Encompass
This manual hand chain hoist provides reliable lifting for various applications. Designed for heavy-duty tasks, it features a substantial capacity and extended lift distance, making it suitable for industrial environments. Its robust construction ensures durability and efficient operation.
  • Manual hand chain hoist type for precise control.
  • 10000 lb lifting capacity for demanding tasks.
  • 20 ft maximum lift distance.
  • Minimum headroom of 23.88 in.
  • Hook mount type for versatile installation.
  • Durable steel chain and hook construction.
  • Equipped with two chains for balanced load distribution.
Catalog No. 50-005-29849 Supplier MSC Supplier No. 06061220
This manual hand chain hoist provides reliable lifting for various applications. Designed for heavy-duty tasks, it features a substantial capacity and extended lift distance, making it suitable for industrial environments. Its robust construction ensures durability and efficient operation.
  • Manual hand chain hoist type for precise control.
  • 10000 lb lifting capacity for demanding tasks.
  • 20 ft maximum lift distance.
  • Minimum headroom of 23.88 in.
  • Hook mount type for versatile installation.
  • Durable steel chain and hook construction.
  • Equipped with two chains for balanced load distribution.