Learn More
MSC CM Manual Hoist, 1500 lb Capacity, 15 in Lift, 12-5/8 in Min Headroom
Catalog No. 5000529198
Encompass
This manual hoist features a 1500 lb capacity and a 12-5/8 in minimum headroom. It is designed for applications requiring precise lifting and controlled handling.
  • 1500 lb capacity
  • 15 in lift height
  • 45 lb pull to lift load
  • Country of origin: China
Catalog No. 50-005-29198 Supplier MSC Supplier No. 18644567
This manual hoist features a 1500 lb capacity and a 12-5/8 in minimum headroom. It is designed for applications requiring precise lifting and controlled handling.
  • 1500 lb capacity
  • 15 in lift height
  • 45 lb pull to lift load
  • Country of origin: China