Learn More
MSC CM Manual Hoist, 3000 lb Capacity, 2 Chain
Catalog No. 5000513481
Encompass
This manual hoist is designed for lifting applications requiring a capacity of 3000 lb. It features a two-chain configuration and requires a lever force of 48 to operate, making it suitable for various industrial and commercial lifting tasks.
  • 3000 lb capacity
  • Two-chain configuration
  • Lever force to operate hoist: 48
  • Backed by a satisfaction guarantee allowing for exchange or refund if expectations are not met, within 30 days of shipment.
Catalog No. 50-005-13481 Supplier MSC Supplier No. 18644658
This manual hoist is designed for lifting applications requiring a capacity of 3000 lb. It features a two-chain configuration and requires a lever force of 48 to operate, making it suitable for various industrial and commercial lifting tasks.
  • 3000 lb capacity
  • Two-chain configuration
  • Lever force to operate hoist: 48
  • Backed by a satisfaction guarantee allowing for exchange or refund if expectations are not met, within 30 days of shipment.