Learn More
MSC CM Manual Lever Hoist, 6000 lb Capacity, 5 ft Max Lift
Catalog No. 5000531913
Encompass
This Manual Lever Hoist offers a 6000 lb capacity and a 5 ft maximum lift distance with a chain length of 5 ft. It features a pull capacity of 95.0 lb and includes overload protection.
  • 6000 lb capacity
  • 5 ft maximum lift distance
  • 5 ft chain length
  • 95.0 lb pull to lift load
  • 17.00 in minimum headroom
  • Lever force to operate hoist is 95 lb
  • 16.94 in distance between hooks
  • Mounts via hook (without trolley)
  • Includes overload protection
  • Steel chain material
  • One chain
Catalog No. 50-005-31913 Supplier MSC Supplier No. 89788889
This Manual Lever Hoist offers a 6000 lb capacity and a 5 ft maximum lift distance with a chain length of 5 ft. It features a pull capacity of 95.0 lb and includes overload protection.
  • 6000 lb capacity
  • 5 ft maximum lift distance
  • 5 ft chain length
  • 95.0 lb pull to lift load
  • 17.00 in minimum headroom
  • Lever force to operate hoist is 95 lb
  • 16.94 in distance between hooks
  • Mounts via hook (without trolley)
  • Includes overload protection
  • Steel chain material
  • One chain