Learn More
Waters Corp Performance Maintenance Kit, ACQUITY CM-S, Annual, Air Filters, Gasket
Catalog No. 502392396 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Performance Maintenance Kit is designed to keep your CM-S instrument operating at peak performance. It contains all the necessary quality parts recommended for annual replacement.
  • Designed for CM-S systems.
  • Ensures instrument operates at peak performance.
  • Recommended for annual replacement.
  • Includes air filters and a gasket.
  • Compatible with Ultra Performance Liquid Chromatography (UPLC) system type.
  • Component for Quaternary Solvent Manager (QSM).
  • One unit per package.
Catalog No. 50-239-2396 Supplier Waters Corp Supplier No. 201000282
This Performance Maintenance Kit is designed to keep your CM-S instrument operating at peak performance. It contains all the necessary quality parts recommended for annual replacement.
  • Designed for CM-S systems.
  • Ensures instrument operates at peak performance.
  • Recommended for annual replacement.
  • Includes air filters and a gasket.
  • Compatible with Ultra Performance Liquid Chromatography (UPLC) system type.
  • Component for Quaternary Solvent Manager (QSM).
  • One unit per package.